Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587699 Supplier Invitrogen™ Supplier No. PA587699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HL-60, MCF7, Jurkat, Raji, Mouse testis, Rat testis; Cellular Location: Membrane, Single-pass type I membrane protein Immunogen sequence: PCGLNQWICI DGVCMSGDKQ CTDTFGKEVE FGPSECYSHL NSKTDVSGNC GISDSGYTQC EADNLQCGKL ICKYVGKFLL QIPRATIIYA NISGHLCIAV EFASDHADSQ KMWIKDGTSC GSNKVCRNQR CVSSSYLGYD CTTDKCNDRG VCNNKKHCHC SASYLPPDCS VQSDLWPGGS IDSGNFPPVA IPARLPERRY IENIYHSKPM R.

ADAM2 is a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. ADAM2 is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions. This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Adam2
Gene Accession No. Q60718, Q63202, Q99965
Gene Alias a disintegrin and metallopeptidase domain 2; a disintegrin and metalloprotease domain 2; a disintegrin and metalloprotease domain (ADAM) 2; a disintegrin and metalloprotease domain 2 (fertilin beta); a disintegrin and metalloproteinase; ADAM; ADAM 2; ADAM metallopeptidase domain 2; Adam2; ADAMs; AI323749; cancer/testis antigen 15; CRYN1; CRYN2; CT15; Disintegrin and metalloproteinase domain-containing protein 2; fertilin beta; fertilin subunit beta; FTNB; metalloendopeptidases; PH30; PH-30; PH-30b; Ph30-beta
Gene Symbols Adam2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 476-686 of human ADAM2 (Q99965).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11495, 2515, 56806
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.