Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572283
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572283 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572283 Supplier Invitrogen™ Supplier No. PA572283
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADAM21 Polyclonal Antibody for Western Blot

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The expression of this gene expression is testis-specific.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM21
Applications Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ADAM21
Gene Accession No. Q9JI76, Q9UKJ8
Gene Alias a disintegrin and metallopeptidase domain 21; a disintegrin and metalloprotease domain (ADAM) 21; a disintegrin and metalloprotease domain 21; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 21; ADAM; ADAM 21; ADAM 31; ADAM metallopeptidase domain 21; ADAM metallopeptidase domain 21, preproprotein; Adam21; Adam31; ADAMs; disintegrin and metalloproteinase domain-containing protein 21; Disintegrin and metalloproteinase domain-containing protein 31; metalloendopeptidases
Gene Symbols ADAM21
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 645-679 amino acids from the C-terminal region of human ADAM21.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56622, 8747
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.