Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530639
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530639 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530639 Supplier Invitrogen™ Supplier No. PA530639
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: IMR32. Predicted reactivity: Human (99%), Mouse (83%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is highly expressed in the brain and may function as an integrin ligand in the brain. Alternative splicing results in several transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM22
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Adam22
Gene Accession No. Q9P0K1, Q9R1V6
Gene Alias 2900022I03Rik; a disintegrin and metallopeptidase domain 22; a disintegrin and metalloprotease domain (ADAM) 22; a disintegrin and metalloprotease domain 22; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 22; ADAM; ADAM 22; ADAM metallopeptidase domain 22; Adam22; ADAMs; AI854032; Disintegrin and metalloproteinase domain-containing protein 22; MDC2; metalloendopeptidases; Metalloproteinase-disintegrin ADAM22-3; metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2; MGC149832
Gene Symbols Adam22
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ADAM22. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11496, 53616
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.