Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569406
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569406 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569406 Supplier Invitrogen™ Supplier No. PA569406
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%.

ADAMs (a disintegrin and metalloproteinase domain), also known as MDCs (metalloproteinase, disintegrin, and cysteine-rich domain) or cellular disintegrins, are a family of proteins that are expressed in numerous different tissues. They catalyze proteolysis, adhesion, fusion and intracellular signaling. ADAMs are membrane-anchored proteins, and there are over 30 different members in the family with many diverse functions. ADAM 23 is exclusively expressed in fetal and adult brain and may function as an integrin ligand in cells of neural origin. Adhesion of neuroblastoma and astrocytoma cells is promoted by the disentegrin-like domain of ADAM23 and is mediated by an interaction of this protein with αvβ3. A short amino acid sequence in the disintegrin loop of ADAM23 interacts with αvβ3 to mediate cell interactions that take place during both normal and pathological processes. The three characteristic histidine residues and the glutamic acid residue typical of metalloproteinases are lacking in the metalloproteinase-like domain of ADAM23. This suggests that ADAM23 may not be involved in protease-mediated events, but could exclusively serve in cell adhesion processes. In addition, evidence suggests that ADAM23 may be a tumor suppressor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM23
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ADAM23
Gene Accession No. Q9R1V7
Gene Alias a disintegrin and metallopeptidase domain 23; a disintegrin and metalloprotease domain 23; a disintegrin and metalloproteinase; ADAM; ADAM 23; ADAM metallopeptidase domain 23; ADAM23; ADAM-23; ADAMs; AW046396; disintegrin and metalloproteinase domain-containing protein 23; MDC3; MDC-3; metalloendopeptidases; Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 3
Gene Symbols ADAM23
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse ADAM23 (aa 773-822).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23792
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.