Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM28 Antibody (1004266), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB93252MFV610
Description
ADAM28 Monoclonal antibody specifically detects ADAM28 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| ADAM28 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 10863 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 1004266 | |
| Flow Cytometry, CyTOF-ready | |
| a disintegrin and metalloproteinase domain 28, ADAM metallopeptidase domain 28, ADAM23MDC-L, disintegrin and metalloproteinase domain-containing protein 28, EC 3.4.24, EC 3.4.24.-, eMDC II, eMDCII, Epididymial metalloproteinase-like, disintegrin-like, and cysteine-rich proteinII, MDCLADAM 28, MDC-Lm, MDC-Ls, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L, metalloproteinase-like, disintegrin-like, and cysteine-rich protein-L | |
| Chinese Hamster Ovary cell line, CHO-derived human ADAM28, Leu72 and Leu191-Ala623,, Accession # Q0UKQ2 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction