Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575835 Supplier Invitrogen™ Supplier No. PA575835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene is a lymphocyte-expressed ADAM protein. Alternative splicing results in two transcript variants. The shorter version encodes a secreted isoform, while the longer version encodes a transmembrane isoform.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM28
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ADAM28
Gene Accession No. Q9JLN6, Q9UKQ2
Gene Alias a disintegrin and metallopeptidase domain 28; a disintegrin and metalloprotease domain 28; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 28; ADAM; ADAM 28; ADAM metallopeptidase domain 28; ADAM23; Adam28; ADAMs; C130072N01Rik; D430033C21Rik; disintegrin 1; disintegrin and metalloproteinase domain-containing protein 28; Dtgn1; eMDC II; eMDCII; Epididymal metalloproteinase-like, disintegrin-like, and cysteine-rich protein II; epididymial metalloproteinase-like, disintegrin-like, and cysteine-rich protein II; MDCL; MDC-L; MDC-Lm; MDC-Ls; metalloendopeptidases; metalloproteinase-disintegrin domain containing protein TECADAM; Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L; metalloproteinase-like, disintegrin-like, and cysteine-rich protein-L; TECADAM; Thymic epithelial cell-ADAM
Gene Symbols ADAM28
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human ADAM28.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10863, 13522, 290344
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.