Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAM29 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550596
Description
The antibody detects endogenous levels of total ADAM29 protein.
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene is highly expressed in testis and may be involved in human spermatogenesis. Alternative splicing results in multiple transcript variants that encode the same protein.
Specifications
ADAM29 | |
Polyclonal | |
Unconjugated | |
ADAM29 | |
a disintegrin and metallopeptidase domain 29; a disintegrin and metalloprotease domain 29; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 29; ADAM; ADAM 29; ADAM metallopeptidase domain 29; ADAM29; ADAMs; Cancer/testis antigen 73; CT73; Disintegrin and metalloproteinase domain-containing protein 29; metallaproteinase-disintegrin (ADAM29); metalloendopeptidases; svph1; testis secretory sperm-binding protein Li 207a | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11086 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9UKF5 | |
ADAM29 | |
Synthetic peptide corresponding to a region derived from internal residues of human ADAM metallopeptidase domain 29. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction