Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569435
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569435 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569435 Supplier Invitrogen™ Supplier No. PA569435
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 85%; Dog: 86%; Guinea Pig: 92%; Horse: 83%; Human: 100%; Mouse: 100%; Rabbit: 91%; Rat: 100%.

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene is highly expressed in testis and may be involved in human spermatogenesis. Alternative splicing results in multiple transcript variants that encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM29
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ADAM29
Gene Accession No. Q811Q4
Gene Alias a disintegrin and metallopeptidase domain 29; a disintegrin and metalloprotease domain 29; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 29; ADAM; ADAM 29; ADAM metallopeptidase domain 29; ADAM29; ADAMs; Cancer/testis antigen 73; CT73; Disintegrin and metalloproteinase domain-containing protein 29; metallaproteinase-disintegrin (ADAM29); metalloendopeptidases; svph1; testis secretory sperm-binding protein Li 207a
Gene Symbols ADAM29
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ADAM29 (aa 101-150).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 244486
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.