Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM30 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP186980
Description
ADAM30 Polyclonal specifically detects ADAM30 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ADAM30 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q9UKF2 | |
ADAM30 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:RCFRKGCRSRYMQCQSIFGPDAMEAPSECYDAVNLIGDQFGNCEITGIRNFKKCESANSICGRLQCINVETIPDLPEHTTIISTHLQAENLMCWGTGYHLSMKPMGIPDLGMINDGTSCG | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
a disintegrin and metalloproteinase domain 30, ADAM 30, ADAM metallopeptidase domain 30, disintegrin and metalloproteinase domain-containing protein 30, EC 3.4.24.-, svph4 | |
Rabbit | |
Affinity Purified | |
RUO | |
11085 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction