Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM30 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169319
Description
ADAM30 Polyclonal specifically detects ADAM30 in Human samples. It is validated for Western Blot.Specifications
ADAM30 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
a disintegrin and metalloproteinase domain 30, ADAM 30, ADAM metallopeptidase domain 30, disintegrin and metalloproteinase domain-containing protein 30, EC 3.4.24.-, svph4 | |
Rabbit | |
66 kDa | |
100 μL | |
Primary | |
Bovine: 85%; Canine: 77%. | |
Human, Bovine, Canine | |
Purified |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9UKF2 | |
ADAM30 | |
Synthetic peptides corresponding to ADAM30(ADAM metallopeptidase domain 30) The peptide sequence was selected from the N terminal of ADAM30. Peptide sequence RLLLPRHLRVFSFTEHGELLEDHPYIPKDCNYMGSVKESLDSKATISTCM. | |
Protein A purified | |
RUO | |
11085 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction