Learn More
Invitrogen™ ADAM30 Monoclonal Antibody (3E5)

Description
Peptide Sequence: YKHPKYLELI LLFDQSRYRF VNNNLSQVIH DAILLTGIMD TYFQDVRMRI HLKALEVWTD FNKIRVGYPE LAEVLGRFVI YKKSVLNARL SSDWAHLYLQ.
Specifications
Specifications
| Antigen | ADAM30 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3E5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ADAM30 |
| Gene Accession No. | Q9UKF2 |
| Gene Alias | a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 30; ADAM; ADAM 30; ADAM metallopeptidase domain 30; ADAM30; ADAMs; Disintegrin and metalloproteinase domain-containing protein 30; metalloendopeptidases; svph4; UNQ2509/PRO5997 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.