Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585442
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585442 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585442 Supplier Invitrogen™ Supplier No. PA585442
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. This product is recommended for use in Proximity Ligation Assay (PLA). Positive Control: Target recombinant protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is testis-specific and contains a polymorphic region, resulting in isoforms with varying numbers of C-terminal repeats.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM30
Applications Proximity Ligation Assay
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ADAM30
Gene Accession No. Q9UKF2
Gene Alias a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 30; ADAM; ADAM 30; ADAM metallopeptidase domain 30; ADAM30; ADAMs; Disintegrin and metalloproteinase domain-containing protein 30; metalloendopeptidases; svph4; UNQ2509/PRO5997
Gene Symbols ADAM30
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human ADAM30. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11085
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.