Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584536
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584536 Supplier Invitrogen™ Supplier No. PA584536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDARCESVFG KGSRNAPFAC YEEIQSQSDR FGNCGRDRNN KYVFCGWRNL ICGRLVCTYP TRKPFHQENG DVIYAF Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 71%.

This gene encodes a member of the disintegrin family of membrane-anchored proteins that play a role in diverse biological processes such as brain development, fertilization, tumor development and inflammation. This gene is predominantly expressed in the testis. The encoded protein undergoes proteolytic processing to generate a mature polypeptide comprised of an metalloprotease, disintegrin and epidermal growth factor-like domains. This gene is located in a cluster of other disintegrin and metallopeptidase family genes on chromosome 8. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Sep 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM32
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADAM32
Gene Accession No. Q8TC27
Gene Alias a disintegrin and metallopeptidase domain 32; a disintegrin and metalloprotease domain 32; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 32; ADAM; ADAM 32; ADAM metallopeptidase domain 32; Adam32; ADAMs; Disintegrin and metalloproteinase domain-containing protein 32; FLJ26299; FLJ29004; metalloendopeptidases; metalloprotease/disintegrin; metalloproteinase 12-like protein; testicular tissue protein Li 13; UNQ5982/PRO21340
Gene Symbols ADAM32
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADAM32. Recombinant protein control fragment (Product #RP-107136).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203102
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.