Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM33 Antibody (508101) [Alexa Fluor™ 532], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5565X
Description
ADAM33 Monoclonal antibody specifically detects ADAM33 in Human samples. It is validated for Immunoprecipitation, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
ADAM33 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Immunoprecipitation, Flow Cytometry (Intracellular Staining), CyTOF | |
508101 | |
Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
a disintegrin and metalloprotease 33, a disintegrin and metalloproteinase domain 33, ADAM 33, ADAM metallopeptidase domain 33, C20orf153, chromosome 20 open reading frame 153, disintegrin and metalloproteinase domain-containing protein 33, disintegrin and reprolysin metalloproteinase family protein, DJ964F7.1, DKFZp434K0521, EC 3.4.24.-, FLJ35308, FLJ36751, MGC149823, MGC71889 | |
Chinese hamster ovary cell line CHO-derived recombinant human ADAM33, Val30-Leu501 (predicted), Accession # Q9BZ11 | |
0.1 mL | |
Asthma, Cell Cycle and Replication, Immunology | |
80332 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction