Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564312
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564312 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564312 Supplier Invitrogen™ Supplier No. PA564312
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSAGDAHGNC GQDSEGHFLP CAGRDALCGK LQCQGGKPSL LAPHMVPVDS TVHLDGQEVT CRGALALPSA QLD Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 64%.

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This protein is a type I transmembrane protein implicated in asthma and bronchial hyperresponsiveness. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM33
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Adam33
Gene Accession No. Q9BZ11
Gene Alias a disintegrin and metallopeptidase domain 33; a disintegrin and metalloprotease 33; a disintegrin and metalloprotease domain 33; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 33; ADAM; ADAM 33; ADAM metallopeptidase domain 33; ADAM33; Adaml; ADAMs; C20orf153; disintegrin and metalloproteinase domain-containing protein 33; disintegrin and reprolysin metalloproteinase family protein; DJ964F7.1; DKFZp434K0521; FLJ35308; FLJ36751; HGNC:15478; metalloendopeptidases; MGC149823; MGC71889; RP23-228H14.4; RP5-964F7.2; UNQ873/PRO1891
Gene Symbols Adam33
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADAM33. Recombinant protein control fragment (Product #RP-103749).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80332
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.