Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544080
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544080 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544080 Supplier Invitrogen™ Supplier No. PA544080
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PTETLGVENK GYFGDEQQIR TEPILPEIHF LNKPASKDSR GIADPNQSAK Sequence homology: Dog: 86%; Horse: 85%; Human: 100%; Mouse: 91%.

The ADAM family is composed of zinc-binding proteins that can function as adhesion proteins and/or endopeptidases. They are involved in a number of biologic processes, including fertilization, neurogenesis, muscle development, and immune response.The ADAM family is composed of zinc-binding proteins that can function as adhesion proteins and/or endopeptidases. They are involved in a number of biologic processes, including fertilization, neurogenesis, muscle development, and immune response.[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ADAM7
Gene Accession No. Q9H2U9
Gene Alias 9230104J12Rik; a disintegrin and metallopeptidase domain 7; a disintegrin and metalloprotease domain (ADAM) 7; a disintegrin and metalloprotease domain 7; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 7; ADAM; ADAM 7; ADAM metallopeptidase domain 7; Adam7; ADAM-7; ADAMs; Disintegrin and metalloproteinase domain-containing protein 7; EAP1; EAPI; epididymal apical protein I; GP83; GP-83; metalloendopeptidases; Sperm maturation-related glycoprotein GP-83
Gene Symbols ADAM7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ADAM7 (aa 705-754).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8756
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.