Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | ADAM8 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ADAM8 Polyclonal specifically detects ADAM8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ADAM8 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
P78325 | |
101 | |
This antibody was developed against a recombinant protein corresponding to amino acids: QYEVVLPWRLPGPRVRRALPSHLGLHPERVSYVLGATGHNFTLHLRKNRDLLGSGYTETYTAANGSEVTEQPRGQDHCFYQGHV | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Polyclonal | |
Rabbit | |
Cellular Markers | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
a disintegrin and metalloproteinase domain 8, ADAM 8, ADAM metallopeptidase domain 8, CD156a antigen, CD156human leukocyte differentiation antigen, Cell surface antigen MS2, EC 3.4.24, EC 3.4.24.-, MGC134985, MS2disintegrin and metalloproteinase domain-containing protein 8 | |
ADAM8 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title