Learn More
ADAM8 Monoclonal Antibody (143303)

Description
ADAM8 Monoclonal Antibody for Western Blot, IP
Specifications
Specifications
| Antigen | ADAM8 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 143303 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | ADAM8 |
| Gene Accession No. | P78325 |
| Gene Alias | a disintegrin and metallopeptidase domain 8; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 8; ADAM; ADAM 8; ADAM metallopeptidase domain 8; Adam8; ADAMs; CD156; CD156a; Cell surface antigen MS2; disintegrin and metalloproteinase domain-containing protein 8; E430039A18Rik; human leukocyte differentiation antigen; Macrophage cysteine-rich glycoprotein; metalloendopeptidases; MGC134985; MS2; RGD1566394 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.