Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA131538
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA131538 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA131538 Supplier Invitrogen™ Supplier No. PA131538
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Recommended positive controls: Human Brain lysate.. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene may be involved in cell adhesion during neurodegeneration, and it is thought to be a target for allergic respiratory diseases, including asthma. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ADAM8
Gene Accession No. P78325
Gene Alias a disintegrin and metallopeptidase domain 8; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 8; ADAM; ADAM 8; ADAM metallopeptidase domain 8; Adam8; ADAMs; CD156; CD156a; Cell surface antigen MS2; disintegrin and metalloproteinase domain-containing protein 8; E430039A18Rik; human leukocyte differentiation antigen; Macrophage cysteine-rich glycoprotein; metalloendopeptidases; MGC134985; MS2; RGD1566394
Gene Symbols ADAM8
Host Species Goat
Immunogen Synthetic peptide: QRKQGAGAPTAP, corresponding to C terminal amino acids 813-824 of ADAM8.
Purification Method IgG fraction
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.