Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM9 Antibody (MM0080-8A15) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | ADAM9 |
---|---|
Clone | MM0080-8A15 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Description
ADAM9 Monoclonal specifically detects ADAM9 in Human samples. It is validated for Western Blot.Specifications
ADAM9 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Signal Transduction | |
Q13443 | |
8754 | |
IgG1 | |
Protein G purified |
MM0080-8A15 | |
Western Blot 1:500-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
a disintegrin and metalloproteinase domain 9 (meltrin gamma), ADAM 9, ADAM metallopeptidase domain 9, ADAM metallopeptidase domain 9 (meltrin gamma), Cellular disintegrin-related protein, cone rod dystrophy 9, CORD9, disintegrin and metalloproteinase domain-containing protein 9, EC 3.4.24, EC 3.4.24.-, MCMPMDC9KIAA0021Mltng, Meltrin-gamma, Metalloprotease/disintegrin/cysteine-rich protein 9, MLTNG, Myeloma cell metalloproteinase | |
ADAM9 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title