Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM9 Antibody (MM0080-8A15), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212021C
Description
ADAM9 Monoclonal specifically detects ADAM9 in Human samples. It is validated for Western Blot.Specifications
ADAM9 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ADAM9 | |
Human recombinant ADAM9 (extracellular domain) | |
0.1 mL | |
Primary | |
Detects human ADAM9 with low cross activity with mouse ADAM9 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
MM0080-8A15 | |
Western Blot | |
a disintegrin and metalloproteinase domain 9 (meltrin gamma), ADAM 9, ADAM metallopeptidase domain 9, ADAM metallopeptidase domain 9 (meltrin gamma), Cellular disintegrin-related protein, cone rod dystrophy 9, CORD9, disintegrin and metalloproteinase domain-containing protein 9, EC 3.4.24, EC 3.4.24.-, MCMPMDC9KIAA0021Mltng, Meltrin-gamma, Metalloprotease/disintegrin/cysteine-rich protein 9, MLTNG, Myeloma cell metalloproteinase | |
Mouse | |
Protein G purified | |
Signal Transduction | |
8754 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction