Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ADAM9 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP159010
![]() |
Novus Biologicals
NBP159010 |
100 μL |
Each for $487.50
|
|
|||||
NBP15901020
![]() |
Novus Biologicals
NBP15901020UL |
20 μL | N/A | N/A | N/A | ||||
Description
ADAM9 Polyclonal specifically detects ADAM9 in Human samples. It is validated for Western Blot.Specifications
ADAM9 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
a disintegrin and metalloproteinase domain 9 (meltrin gamma), ADAM 9, ADAM metallopeptidase domain 9, ADAM metallopeptidase domain 9 (meltrin gamma), Cellular disintegrin-related protein, cone rod dystrophy 9, CORD9, disintegrin and metalloproteinase domain-containing protein 9, EC 3.4.24, EC 3.4.24.-, MCMPMDC9KIAA0021Mltng, Meltrin-gamma, Metalloprotease/disintegrin/cysteine-rich protein 9, MLTNG, Myeloma cell metalloproteinase | |
ADAM9 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
B7ZLN7 | |
8754 | |
Synthetic peptides corresponding to ADAM9(ADAM metallopeptidase domain 9 (meltrin gamma)) The peptide sequence was selected from the middle region of ADAM9. Peptide sequence NKGGNCLLNIPKPDEAYSAPSCGNKLVDAGEECDCGTPKECELDPCCEGS. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title