Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adamantane 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
1kg
Packaging:
Glass bottle
281-23-2
C10H16
136.24
MFCD00074719
ORILYTVJVMAKLC-UHFFFAOYSA-N
tricyclo 3.3.1.13,7 decane,adamantan,tricyclo 3.3.1.1 3,7 decane,tricyclo 3.3.1.1 decane,unii-pjy633525u,tricyclo 3.3.1.1~3,7~ decane,adamantine,2-adamantyl,adamantan-1-yl
9238
CHEBI:40519
adamantane
C1C2CC3CC1CC(C2)C3
281-23-2
C10H16
136.24
MFCD00074719
ORILYTVJVMAKLC-UHFFFAOYSA-N
tricyclo 3.3.1.13,7 decane,adamantan,tricyclo 3.3.1.1 3,7 decane,tricyclo 3.3.1.1 decane,unii-pjy633525u,tricyclo 3.3.1.1~3,7~ decane,adamantine,2-adamantyl,adamantan-1-yl
9238
CHEBI:40519
adamantane
C1C2CC3CC1CC(C2)C3
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Melting Point 205.0°C to 210.0°C
Color White to Beige
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Merck Index 15,137
Solubility Information Solubility in water: insoluble. Other solubilities: 8% in light petroleum fract. (20°C),6.7% in diethyl ether-1.7% in ethyl acetate (20°C),9.1% in methylene chloride-0.8% in acetone (20°C),10% in chloroform-12.5% in ccl4 (20°C),0.5%
Formula Weight 136.24
Physical Form Crystals or Crystalline Powder
Percent Purity ≥99%
Chemical Name or Material Adamantane, 99+%
Show More Show Less

EINECSNumber : 206-001-4

RTECSNumber : YK0493000

TSCA : TSCA