Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMDEC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103574 Supplier Invitrogen™ Supplier No. PA5103574
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ADAMDEC1.

ADAMDEC1 may play an important role in the control of the immune response and during pregnancy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMDEC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADAMDEC1
Gene Accession No. O15204, Q9R0X2
Gene Alias 2210414L24Rik; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain-like protein decysin 1; a disintegrin and metalloproteinase domain-like protein decysin-1; ADAM; ADAM DEC1; ADAM like decysin 1; ADAMDEC1; ADAM-like protein decysin 1; ADAM-like protein decysin-1; ADAM-like, decysin 1; ADAMs; AI574231; Dcsn; decysin; disintegrin metalloprotease (decysin); disintegrin protease; M12.219; metalloendopeptidases; Unknown (protein for IMAGE:7929269)
Gene Symbols ADAMDEC1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADAMDEC1(Accession O15204), corresponding to amino acid residues G437-E470.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27299, 58860
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.