Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS1 Antibody (1G5D5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | ADAMTS1 |
---|---|
Clone | 1G5D5 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61808
![]() |
Novus Biologicals
NBP261808 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
ADAMTS1 Monoclonal specifically detects ADAMTS1 in Human samples. It is validated for Western Blot, ELISA.Specifications
ADAMTS1 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 1, ADAM metallopeptidase with thrombospondin type 1 motif, 1, ADAM-TS 1, ADAM-TS1, ADAMTS-1, C3-C5, METH-1 | |
ADAMTS1 | |
IgG1 | |
Protein G purified |
1G5D5 | |
Western Blot 1:500-1:2000, ELISA 1:10000 | |
Monoclonal | |
Purified | |
Cell Cycle and Replication | |
9510 | |
Purified recombinant fragment of human ADAMTS1 (AA: 858-960) expressed in E. Coli. | |
Primary | |
147 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title