Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS1 Antibody (OTI4H2), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271474UV
Description
ADAMTS1 Monoclonal specifically detects ADAMTS1 in Human samples. It is validated for Western Blot.Specifications
ADAMTS1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ADAMTS1 | |
Recombinant protein fragment corresponding to amino acids 412-644 of human ADAMTS1 (NP_008919) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot | |
OTI4H2 | |
Western Blot | |
a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 1, ADAM metallopeptidase with thrombospondin type 1 motif, 1, ADAM-TS 1, ADAM-TS1, ADAMTS-1, C3-C5, METH-1 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
9510.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction