Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS1 Antibody (OTI4H2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246492
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
ADAMTS1 Monoclonal specifically detects ADAMTS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ADAMTS1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 1, ADAM metallopeptidase with thrombospondin type 1 motif, 1, ADAM-TS 1, ADAM-TS1, ADAMTS-1, C3-C5, METH-1 | |
Mouse | |
100.3 kDa | |
0.1 mL | |
Cell Cycle and Replication | |
9510 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI4H2 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
ADAMTS1 | |
Human recombinant protein fragment corresponding to amino acids 412-644 of human ADAMTS1 (NP_008919) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction