Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS10 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299535JF646
Description
ADAMTS10 Polyclonal antibody specifically detects ADAMTS10 in Mouse samples. It is validated for Western BlotSpecifications
ADAMTS10 | |
Polyclonal | |
Western Blot | |
a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 10, ADAM metallopeptidase with thrombospondin type 1 motif, 10, ADAM-TS 10, ADAMTS-10, ADAM-TS10A disintegrin and metalloproteinase with thrombospondin motifs 10, EC 3.4.24.-, EC 3.4.24.82, WMS, zinc metalloendopeptidase | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ADAMTS10 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
81794 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction