Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112124
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112124 Supplier Invitrogen™ Supplier No. PA5112124
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADAMTS10 Polyclonal Antibody for Western Blot

ADAMTS (a disintegrin and metalloproteinase domain with Thrombospondin type-1 modules) is a family of zinc-dependent proteases that are implicated in a variety of normal and pathological conditions, including arthritis and cancer (1-3). Embryogenesis, morphological growth changes, inflammation, tumor invasion and metastasis involve the breakdown and remodeling of the extracellular matrix. This degradation is due to the family of enzymes known as the matrix metalloproteinases (MMPs), which are related to ADAMTS subtypes. ADAMTS protein family members contain an amino-terminal propeptide domain, a metalloproteinase domain, a disintegrin-like domain and a carboxy-terminus that contains a varying number of thrombospondin type-1 (TSP-1) motifs (1-3). ADAMTS genes are primarily expressed in fetal tissues, including the lung, kidney and liver (1-3). The human ADAMTS10 gene maps to chromosome 19p13.3 (4). The human ADAMTS19 gene maps to chromosome 5q31 (1,5).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ADAMTS10
Gene Accession No. P58459
Gene Alias 9430006O18; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 10; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 10; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 10; a disintegrin-like and metalloprotease with thrombospondin type 1 motif, 10; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 10; ADAM metallopeptidase with thrombospondin type 1 motif, 10; ADAMs; ADAM-TS 10; ADAMTS10; Adam-ts10; ADAMTS-10; AW045948; metalloendopeptidases; WMS; WMS1; zinc metalloendopeptidase; Znmp
Gene Symbols ADAMTS10
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Mouse ADAMTS10.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224697
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.