Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAMTS12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103576
Description
Antibody detects endogenous levels of total ADAMTS12.
This protein is a disintegrin and metalloproteinase with thrombospondin motifs-12 (ADAMTS12), which is a member of the ADAMTS protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS-1) motif. Individual members of this family differ in the number of C-terminal TS-1 motifs, and some have unique C-terminal domains. The enzyme encoded by this gene contains eight TS-1 motifs. It may play roles in pulmonary cells during fetal development or in tumor processes through its proteolytic activity or as a molecule potentially involved in regulation of cell adhesion.
Specifications
ADAMTS12 | |
Polyclonal | |
Unconjugated | |
ADAMTS12 | |
a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 12; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 12; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 12; a disintegrin-like and metalloprotease with thrombospondin type 1 motif, 12; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 12; ADAM metallopeptidase with thrombospondin type 1 motif, 12; ADAMs; ADAM-TS 12; ADAMTS12; ADAM-TS12; ADAMTS-12; AI605170; metalloendopeptidases; metalloprotease disintegrin 12; PRO4389; UNQ1918; UNQ1918/PRO4389 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
81792 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P58397 | |
ADAMTS12 | |
A synthesized peptide derived from human ADAMTS12(Accession P58397), corresponding to amino acid residues T1141-S1191. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction