Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS13 Antibody, DyLight 350, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10055261UV
Description
ADAMTS13 Polyclonal antibody specifically detects ADAMTS13 in Human samples. It is validated for Western BlotSpecifications
ADAMTS13 | |
Polyclonal | |
Western Blot | |
A disintegrin and metalloproteinase with thrombospondin motifs 13, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 13, ADAM metallopeptidase with thrombospondin type 1 motif, 13, ADAM-TS13, ADAMTS-13, DKFZp434C2322, EC 3.4.24.14, EC 3.4.24.82, EC 3.4.24.87, FLJ42993, MGC118899, MGC118900, TTPADAM-TS 13, vWF-cleaving protease, vWF-CPC9orf8, VWFCPvon Willebrand factor-cleaving protease | |
The immunogen recognized by this antibody maps to a region between residue 825 and 875 of human ADAM (a disintegrin-like and metalloprotease) metallopeptidase with thrombospondin type 1 motif, 13 using the numbering given in entry NP_620594.1 (GeneID 1109 | |
0.1 mL | |
Cancer, Signal Transduction | |
11093 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction