Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS13 Mouse anti-Human, Alexa Fluor 488, Clone: 442323, R&D Systems™

Catalog No. IC4245G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4245G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4245G100 Supplier R&D Systems Supplier No. IC4245G100UG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADAMTS13 Monoclonal specifically detects ADAMTS13 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen ADAMTS13
Applications Flow Cytometry
Classification Monoclonal
Clone 442323
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q76LX8
Gene Alias A disintegrin and metalloproteinase with thrombospondin motifs 13, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 13, ADAM metallopeptidase with thrombospondin type 1 motif, 13, ADAM-TS13, ADAMTS-13, DKFZp434C2322, EC 3.4.24.14, EC 3.4.24.82, EC 3.4.24.87, FLJ42993, MGC118899, MGC118900, TTP, TTPADAM-TS 13, vWF-cleaving protease, vWF-CP, vWF-CPC9orf8, VWFCPvon Willebrand factor-cleaving protease
Gene Symbols ADAMTS13
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ADAMTS13 isoform 1 Gln34-Trp688 (predicted) Accession # Q76LX8
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11093
Test Specificity Detects human ADAMTS13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ADAMTS1, rhADAMTS4, rhADAMTS5, or rhADAMTS-L1.2 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.