Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103577
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103577 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103577 Supplier Invitrogen™ Supplier No. PA5103577
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ADAMTS13.

ADAMTS13 is a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme is the von Willebrand Factor (vWF)-cleaving protease, which is responsible for cleaving at the site of Tyr842-Met843 of the vWF molecule. A deficiency of this enzyme is associated with thrombotic thrombocytopenic purpura.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS13
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADAMTS13
Gene Accession No. Q76LX8
Gene Alias a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 13; A disintegrin and metalloproteinase with thrombospondin motifs 13-like; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 13; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 13; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 13; ADAM metallopeptidase with thrombospondin type 1 motif, 13; ADAMs; ADAM-TS 13; ADAMTS13; ADAM-TS13; ADAMTS-13; C9orf8; Gm710; LOC102554393; metalloendopeptidases; UNQ6102/PRO20085; von Willebrand factor-cleaving protease; vWF-cleaving protease; VWFCP; vWF-CP; vWF-CP mRNA for von Willebrand factor-cleaving
Gene Symbols ADAMTS13
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADAMTS13(Accession Q76LX8), corresponding to amino acid residues S1162-D1212.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11093
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.