Learn More
ADAMTS13 Goat anti-Human, Polyclonal, Bethyl Laboratories
Goat Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301003A
Description
The recommended shelf life for this product is 1 year from date of receipt.
ADAMTS13 is a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme is the von Willebrand Factor (vWF)-cleaving protease, which is responsible for cleaving at the site of Tyr842-Met843 of the vWF molecule. A deficiency of this enzyme is associated with thrombotic thrombocytopenic purpura.Specifications
| ADAMTS13 | |
| Polyclonal | |
| Unconjugated | |
| ADAMTS13 | |
| von Willebrand factor-cleaving protease; vWF-cleaving protease; vWF-CP; A disintegrin and metalloproteinase with thrombospondin motifs 13; ADAMTS-13; ADAM-TS 13; ADAM-TS13; ADAMTS13; C9orf8 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 11093 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q76LX8 | |
| ADAMTS13 | |
| Between 1377 and C-term | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.