Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS14 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550597

Catalog No. PA550597


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total ADAMTS14 protein.

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. This gene is highly similar to two family members, ADAMTS2 and ADAMTS3, in its sequence and gene structure, and the encoded protein shares the aminoprocollagen peptidase activity with the protein products encoded by ADAMTS2 and ADAMTS3. Various transcript variants of this gene have been identified. They result from the use of two different promoters and transcription initiation sites as well as alternative splicing sites. The full length nature of some transcripts has not been defined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ADAMTS14
Polyclonal
Unconjugated
ADAMTS14
a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 14; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 14; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 14; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 14; ADAM metallopeptidase with thrombospondin type 1 motif, 14; ADAMs; ADAM-TS 14; ADAMTS14; ADAM-TS14; Adamts-14; ATS14; FLJ32820; metalloendopeptidases; TS14
Rabbit
Antigen affinity chromatography
RUO
140766
-20°C
Liquid
Immunohistochemistry (Paraffin)
2.1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q8WXS8
ADAMTS14
Synthetic peptide corresponding to a region derived from internal residues of human ADAM metallopeptidase with thrombospondin type 1 motif, 14.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.