Learn More
Invitrogen™ ADAMTS15 Monoclonal Antibody (561819)

Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | ADAMTS15 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 561819 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | ADAMTS15 |
| Gene Accession No. | Q8TE58 |
| Gene Alias | a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 15; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 15; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 15; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 15, preproprotein; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 15; ADAM metallopeptidase with thrombospondin type 1 motif, 15; ADAMs; ADAM-TS 15; Adamts15; ADAM-TS15; ADAMTS-15; metalloendopeptidases; metalloprotease disintegrin 15 with thrombospondin domains |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.