Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS15 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA548070
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548070 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548070 Supplier Thermo Scientific Supplier No. PA548070

Sheep Polyclonal Antibody

In direct ELISAs, less than 1% cross-reactivity with recombinant human ADAMTS1, L1.2, L2, 3, 4, 5, 8, 10, 12, 13, and 16 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

ADAMTS15 Polyclonal antibody specifically detects ADAMTS15 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS15
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ADAMTS15
Gene Accession No. P59384, Q8TE58
Gene Alias a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 15; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 15; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 15; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 15, preproprotein; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 15; ADAM metallopeptidase with thrombospondin type 1 motif, 15; ADAMs; ADAM-TS 15; Adamts15; ADAM-TS15; ADAMTS-15; metalloendopeptidases; metalloprotease disintegrin 15 with thrombospondin domains
Gene Symbols ADAMTS15
Host Species Sheep
Immunogen CHO-derived recombinant human ADAMTS15 Gly18-Cys682
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170689, 235130
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.