Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS15 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS5857R

Description
ADAMTS proteases are secreted enzymes containing a prometalloprotease domain of the reprolysin type. The ADAMTS proteases function in processing of procollagens and von Willebrand factor as well as catabolism of aggrecan, versican and brevican. They have been demonstrated to have important roles in connective tissue organization, coagulation, inflammation, arthritis, angiogenesis and cell migration. The immunizing peptide sequence is 100% conserved between human, mouse and rat.Specifications
ADAMTS15 | |
Polyclonal | |
Unconjugated | |
ADAMTS15 | |
a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 15; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 15; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 15; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 15, preproprotein; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 15; ADAMs; ADAM-TS 15; Adamts15; ADAM-TS15; ADAMTS-15; metalloendopeptidases; metalloprotease disintegrin 15 with thrombospondin domains | |
Rabbit | |
Protein A | |
RUO | |
170689 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8TE58 | |
ADAMTS15 | |
KLH conjugated synthetic peptide derived from human ADAMTS15. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction