Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS16 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$249.00 - $536.00
Specifications
| Antigen | ADAMTS16 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIPA5122286
|
Invitrogen™
PA5122286 |
20 μL |
Each for $249.00
|
|
|||||
PIPA5122285
![]() |
Invitrogen™
PA5122285 |
100 μL |
Each for $536.00
|
|
|||||
Description
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has high sequence similarity to the protein encoded by ADAMTS18, another family member.Specifications
| ADAMTS16 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8TE57 | |
| 170690 | |
| Leu290-Pro641 (Accession Q8TE57), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ADAMTS16 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 16; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 16; a disintegrin-like and metallopetidase (reprolysin type) with thrombospondin type 1 motif, 16; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 16; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 16 preproprotein; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 16; ADAM metallopeptidase with thrombospondin type 1 motif, 16; ADAMs; ADAM-TS 16; ADAMTS16; ADAM-TS16; ADAMTS-16; ADAMTS16s; KIAA2029; metalloendopeptidases | |
| ADAMTS16 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title