Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS17 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | ADAMTS17 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ADAMTS17 Polyclonal specifically detects ADAMTS17 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ADAMTS17 | |
Polyclonal | |
Rabbit | |
Human | |
A disintegrin and metalloproteinase with thrombospondin motifs 17, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 17, ADAM metallopeptidase with thrombospondin type 1 motif, 17, ADAM-TS 17, ADAM-TS17, ADAMTS-17, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, EC 3.4.24.82, FLJ16363, FLJ32769 | |
ADAMTS17 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
170691 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QVRRCNLHPCQSRWVAGPWSPCSATCEKGFQHREVTCVYQLQNGTHVATRPLYCPGPRPAAVQSCEGQDCLSIWEASEWS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title