Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103580
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103580 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103580 Supplier Invitrogen™ Supplier No. PA5103580
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ADAMTS17.

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The encoded preproprotein is proteolytically processed to generate the mature protein, which may promote breast cancer cell growth and survival. Mutations in this gene are associated with a Weill-Marchesani-like syndrome, which is characterized by lenticular myopia, ectopia lentis, glaucoma, spherophakia, and short stature. [provided by RefSeq, May 2016].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADAMTS17
Gene Accession No. Q8TE56
Gene Alias a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 17; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 17; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 17; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 17; ADAM metallopeptidase with thrombospondin type 1 motif, 17; ADAMs; ADAM-TS 17; ADAMTS17; ADAM-TS17; ADAMTS-17; ATS17; AU023434; disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 17; FLJ16363; FLJ32769; metalloendopeptidases
Gene Symbols ADAMTS17
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADAMTS17(Accession Q8TE56), corresponding to amino acid residues R821-S871.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170691, 233332, 293004
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.