Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560675
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560675 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560675 Supplier Invitrogen™ Supplier No. PA560675
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VFVTPVEVDS AGSYISHDIL HNGRKKRSAQ NARSSLHYRF SAFGQELHLE LKPSAILSSH FIVQVLGKDG ASETQKPEVQ QCFYQGFIRN DSSSSVAVST Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has a high sequence similarity to the protein encoded by gene ADAMTS16, another family member. It is thought to function as a tumor suppressor. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS18
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADAMTS18
Gene Accession No. Q8TE60
Gene Alias 9630038L21; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 18; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 18; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 18; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 21; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 18; ADAM metallopeptidase with thrombospondin type 1 motif, 18; ADAMs; ADAM-TS 18; ADAMTS18; ADAM-TS18; ADAMTS-18; ADAMTS21; disintegrin and metalloprotease-like protein; E130314N14Rik; KNO2; metalloendopeptidases; MMCAT; RGD1560118; sb:eu2468
Gene Symbols ADAMTS18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADAMTS18. Recombinant protein control fragment (Product #RP-92473).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170692
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.