Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563256
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563256 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563256 Supplier Invitrogen™ Supplier No. PA563256
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPLNDTMAIT GHPHRVYRQK RSMEEKVTEK SALHSHYCGI ISDKGRPRSR KIAESGRGKR YSYKLPQEYN IETVVVADPA MVSYHGAD Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

This gene encodes a member of the ADAMTS protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has high sequence similarity to the protein encoded by ADAMTS16, another family member.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS19
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADAMTS19
Gene Accession No. Q8TE59
Gene Alias 4831407I23Rik; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 19; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 19; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 19; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 19; ADAM metallopeptidase with thrombospondin type 1 motif, 19; ADAMs; ADAM-TS 19; Adamts19; ADAM-TS19; ADAMTS-19; AU015154; D230034E10Rik; metalloendopeptidases
Gene Symbols ADAMTS19
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADAMTS19. Recombinant protein control fragment (Product #RP-93162).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171019
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.