Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAMTS19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566170
Description
Immunogen sequence: DSEARDCNGPR KQYRICENPP CPAGLPGFRD WQCQAYSVRT SSPKHILQWQ AVLDEEKPCA LFCSPVGKEQ PILLSEKVMD GTSCGYQ Highest antigen sequence identity to the following orthologs - mouse 94%, rat 82%.
This gene encodes a member of the ADAMTS protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has high sequence similarity to the protein encoded by ADAMTS16, another family member.
Specifications
ADAMTS19 | |
Polyclonal | |
Unconjugated | |
ADAMTS19 | |
4831407I23Rik; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 19; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 19; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 19; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 19; ADAM metallopeptidase with thrombospondin type 1 motif, 19; ADAMs; ADAM-TS 19; Adamts19; ADAM-TS19; ADAMTS-19; AU015154; D230034E10Rik; metalloendopeptidases | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171019 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TE59 | |
ADAMTS19 | |
Recombinant Human ADAMTS19. Recombinant protein control fragment (Product #RP-106856). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction