Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555855
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555855 Supplier Invitrogen™ Supplier No. PA555855
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VAMEVRPSPS TPLEVPLNAS STNATEDHPE TNAVDEPYKI HGLEDEVQPS NLIPRRPSPY EKTRNQRIQE LIDEMRKKEM LG Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 71%.

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene excises the N-propeptide of type I, type II and type V procollagens. Mutations in this gene cause Ehlers-Danlos syndrome type VIIC, a recessively inherited connective-tissue disorder. Alternative splicing results in two transcript variants. The short transcript encodes a protein which has no significant procollagen N-peptidase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Adamts2
Gene Accession No. O95450
Gene Alias a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 2; a disintegrin and metalloproteinase with thrombospondin repeats; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 2; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 2; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 2; ADAM metallopeptidase with thrombospondin type 1 motif, 2; ADAMs; ADAM-TS 2; ADAMTS2; ADAM-TS2; ADAMTS-2; ADAMTS-3; hPCPNI; metalloendopeptidases; mKIAA4060; NPI; PC I-NP; PCINP; PCI-NP; PCPNI; PNPI; Procollagen I N-proteinase; procollagen I/II amino propeptide-processing enzyme; Procollagen N-endopeptidase; procollagen N-proteinase; RGD1565950
Gene Symbols Adamts2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADAMTS2. Recombinant protein control fragment (Product #RP-93160).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9509
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.