Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAMTS20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568085
Description
ADAMTS20 Polyclonal Antibody for Western Blot
May play a role in tissue-remodeling process occurring in both normal and pathological conditions. May have a protease-independent function in the transport from the endoplasmic reticulum to the Golgi apparatus of secretory cargos, mediated by the GON domain.
Specifications
ADAMTS20 | |
Polyclonal | |
Unconjugated | |
ADAMTS20 | |
a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 20; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 20; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 20; ADAM metallopeptidase with thrombospondin type 1 motif, 20; ADAMs; ADAM-TS 20; ADAMTS20; ADAM-TS20; ADAMTS-20; GON-1; metalloendopeptidases | |
Rabbit | |
Affinity Chromatography | |
RUO | |
80070 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P59510 | |
ADAMTS20 | |
Synthesized peptide derived from human ADAMTS20 within amino acids 1-35 of Uniprot ID P59510. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction