Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103582
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103582 Supplier Invitrogen™ Supplier No. PA5103582
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The immunogen for PA5-103582 is CLKDERRHSR KKRLI. Antibody detects endogenous levels of total ADAMTS20.

May play a role in tissue-remodeling process occurring in both normal and pathological conditions. May have a protease-independent function in the transport from the endoplasmic reticulum to the Golgi apparatus of secretory cargos, mediated by the GON domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS20
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADAMTS20
Gene Accession No. P59510, P59511
Gene Alias a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 20; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 20; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 20; ADAM metallopeptidase with thrombospondin type 1 motif, 20; ADAMs; ADAM-TS 20; ADAMTS20; ADAM-TS20; ADAMTS-20; GON-1; metalloendopeptidases
Gene Symbols ADAMTS20
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADAMTS20(Accession P59510), corresponding to amino acid residues N222-V272.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223838, 80070
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.