Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAMTS20 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572343
Description
May play a role in tissue-remodeling process occurring in both normal and pathological conditions. May have a protease-independent function in the transport from the endoplasmic reticulum to the Golgi apparatus of secretory cargos, mediated by the GON domain.
Specifications
ADAMTS20 | |
Polyclonal | |
Unconjugated | |
ADAMTS20 | |
A disintegrin and metalloproteinase with thrombospondin motifs 20; ADAM-TS 20; ADAM-TS20; ADAMTS-20; 3.4.24.-; ADAMTS20 | |
Rabbit | |
Protein A | |
RUO | |
80070 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.47 mg/mL | |
PBS with 0.09% sodium azide | |
P59510 | |
ADAMTS20 | |
KLH conjugated synthetic peptide between 782-816 amino acids from the Central region of human ADAMTS20 | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction