Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAMTS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554390 Supplier Invitrogen™ Supplier No. PA554390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPDGANLRQR SAQQAGSKTV RLVTVPSSPP TKRVHLSSAS QMAAASFFAA SDSIGASSQA RTSKKDGKII D Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 30%.

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene is the major procollagen II N-propeptidase. A deficiency of this protein may be responsible for dermatosparaxis, a genetic defect of connective tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADAMTS3
Gene Accession No. O15072
Gene Alias a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 3; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 3; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 3; ADAM metallopeptidase with thrombospondin type 1 motif, 3; ADAMs; ADAM-TS 3; ADAMTS3; ADAM-TS3; ADAMTS-3; ADAMTS-4; KIAA0366; metalloendopeptidases; PC II-NP; procollagen II amino propeptide-processing enzyme; Procollagen II N-proteinase; zinc metalloendopeptidase
Gene Symbols ADAMTS3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADAMTS3. Recombinant protein control fragment (Product #RP-93163).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9508
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.