Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAMTS3 Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547744
Description
In direct ELISAs and Western blots, approximately 3% cross-reactivity with recombinant human (rh) ADAMTS4 and less than 1% cross-reactivity with rhADAMTS1, rhADAMTS5, rhADAMTS13, and rhADAMTSL1.2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene is the major procollagen II N-propeptidase. A deficiency of this protein may be responsible for dermatosparaxis, a genetic defect of connective tissues.
Specifications
ADAMTS3 | |
Polyclonal | |
Unconjugated | |
ADAMTS3 | |
a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 3; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 3; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 3; ADAM metallopeptidase with thrombospondin type 1 motif, 3; ADAMs; ADAM-TS 3; ADAMTS3; ADAM-TS3; ADAMTS-3; ADAMTS-4; KIAA0366; metalloendopeptidases; PC II-NP; procollagen II amino propeptide-processing enzyme; Procollagen II N-proteinase; zinc metalloendopeptidase | |
Sheep | |
Antigen Affinity Chromatography | |
RUO | |
9508 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O15072 | |
ADAMTS3 | |
CHO-derived recombinant human ADAMTS3 Ser19-Cys712. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction